Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008235.2 | NP_032261.1 | 4 | 1244 | ||
GenBank mRNA | AK132536.1 | - | 4 | 1256 | ||
AK133538.1 | - | 4 | 1260 | |||
BC018375.1 | - | 4 | 1244 | 4505427 | ||
BC051428.1 | - | 4 | 1245 | 6478994 |