Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfb2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008249.4 | NP_032275.2 | 4 | 742 | ||
GenBank mRNA | AK090106.1 | - | 4 | 713 | ||
AK146563.1 | - | 4 | 743 | |||
AK166717.1 | - | 4 | 729 | |||
AK167930.1 | - | 4 | 694 | |||
AK168970.1 | - | 4 | 721 | |||
AK169138.1 | - | 4 | 721 | |||
AK185078.1 | - | 3 | 301 | |||
AK210035.1 | - | 3 | 307 | |||
BC138836.1 | - | 4 | 687 | 8861858 | ||
BC138837.1 | - | 4 | 687 | 8861859 | ||
M74555.1 | - | 4 | 693 |