Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005191.3 | NP_005182.1 | 2, 3 | 503 | ||
XM_017007520.1 | XP_016863009.1 | 2, 3 | 578 | |||
GenBank mRNA | AK292129.1 | - | 2, 3 | 487 | ||
AY081815.1 | - | 1, 2 | 219 | |||
AY197777.1 | - | 1, 2 | 108 | |||
AY197778.1 | - | 1, 2 | 108 | |||
BC042665.1 | - | 2, 3 | 471 | 5181343 | ||
BP225173.1 | - | 2, 3 | 530 | |||
M27533.1 | - | 2, 3 | 425 |