Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008256.4 | NP_032282.2 | 4 | 772 | ||
GenBank mRNA | AK004865.1 | - | 4 | 770 | ||
AK004902.1 | - | 4 | 768 | |||
AK098104.1 | - | 4 | 768 | |||
BC014714.1 | - | 4 | 758 | 4216637 | ||
BC024744.1 | - | 4 | 756 | 5123821 |