Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008258.1 | NP_032284.1 | 5 | 634 | ||
GenBank mRNA | AK004909.1 | - | 5 | 648 | ||
AK010057.1 | - | 5 | 649 | |||
AK013122.1 | - | 5 | 646 | |||
AK014212.1 | - | 5 | 650 | |||
AK150410.1 | - | 5 | 653 | |||
AK152560.1 | - | 5 | 651 | |||
AK159784.1 | - | 5 | 648 | |||
AK167466.1 | - | 5 | 648 | |||
AK167961.1 | - | 5 | 638 | |||
AK168632.1 | - | 5 | 649 | |||
BC003758.1 | - | 5 | 623 | 3591830 | ||
U90123.1 | - | 5 | 634 |