Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008258.1 | NP_032284.1 | 5 | 1061 | ||
GenBank mRNA | AK004909.1 | - | 5 | 1075 | ||
AK010057.1 | - | 5 | 1076 | |||
AK013122.1 | - | 5 | 1073 | |||
AK014212.1 | - | 5 | 1075 | |||
AK150410.1 | - | 5 | 1080 | |||
AK152560.1 | - | 5 | 1078 | |||
AK159784.1 | - | 6 | 992 | |||
AK167961.1 | - | 5 | 1065 | |||
AK168632.1 | - | 5 | 1076 | |||
BC003758.1 | - | 5 | 1050 | 3591830 | ||
U90123.1 | - | 5 | 1061 |