Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008270.2 | NP_032296.2 | 2 | 1729 | ||
GenBank mRNA | AK078860.1 | - | 1 | 1128 | ||
M34857.1 | - | 2 | 1147 |