Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122752.1 | NP_001116224.1 | 2 | 402 | ||
NM_005025.4 | NP_005016.1 | 2 | 635 | |||
XM_017006618.1 | XP_016862107.1 | 2 | 700 | |||
GenBank mRNA | AK290082.1 | - | 2 | 325 | ||
CR627434.1 | - | 2 | 624 | |||
DA359846.1 | - | 2 | 402 | |||
KU178259.1 | - | 1 | 204 | |||
KU178260.1 | - | 1 | 204 | |||
Z81326.1 | - | 2 | 285 |