Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008278.2 | NP_032304.2 | 7 | 1337 | ||
GenBank mRNA | AK002472.1 | - | 7 | 1297 | ||
AK146038.1 | - | 7 | 1305 | |||
BC021157.1 | - | 7 | 1296 | 4208980 | ||
BY581851.1 | - | 1 | 23 | |||
U44389.1 | - | 7 | 1291 |