Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044751.1 | NP_001038216.1 | 5 | 637 | ||
NM_008288.2 | NP_032314.2 | 6 | 663 | |||
GenBank mRNA | AK149518.1 | - | 5 | 635 | ||
AK165212.1 | - | 5 | 635 | |||
AK167419.1 | - | 4 | 587 | |||
AK202781.1 | - | 2 | 99 | |||
BC132364.1 | - | 6 | 603 | 40130641 | ||
BC138780.1 | - | 6 | 603 | 8861802 | ||
BC145415.1 | - | 6 | 603 | 9053973 | ||
DQ089001.1 | - | 5 | 539 | |||
S75207.1 | - | 5 | 584 | |||
X83202.1 | - | 6 | 666 |