Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008296.2 | NP_032322.1 | 4 | 548 | ||
GenBank mRNA | BC013716.1 | - | 4 | 548 | 3978881 | |
BC094064.1 | - | 4 | 558 | 4158683 | ||
KM043052.1 | - | 4 | 548 | |||
KM043053.1 | - | 4 | 548 | |||
KM043054.1 | - | 4 | 548 | |||
KM043055.1 | - | 4 | 548 | |||
X61753.1 | - | 4 | 531 |