Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008296.2 | NP_032322.1 | 8 | 951 | ||
GenBank mRNA | AK075651.1 | - | 4 | 726 | ||
AK089144.1 | - | 7 | 854 | |||
BC013716.1 | - | 8 | 951 | 3978881 | ||
BC094064.1 | - | 8 | 961 | 4158683 | ||
KM043052.1 | - | 8 | 951 | |||
KM043053.1 | - | 8 | 951 | |||
KM043054.1 | - | 8 | 951 | |||
KM043055.1 | - | 8 | 951 | |||
X61753.1 | - | 8 | 934 |