Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164671.1 | NP_001158143.1 | 5 | 702 | ||
NM_001164672.1 | NP_001158144.1 | 5 | 559 | |||
NM_008298.5 | NP_032324.1 | 5 | 653 | |||
GenBank mRNA | AF055664.1 | - | 5 | 568 | ||
AK049963.1 | - | 5 | 639 | |||
AK083046.1 | - | 5 | 589 | |||
AK145963.1 | - | 5 | 635 | |||
AK147604.1 | - | 1 | 3447 | |||
AK167139.1 | - | 5 | 589 | |||
AK167212.1 | - | 5 | 589 | |||
BC057876.1 | - | 5 | 564 | 5347227 | ||
BC060653.1 | - | 4 | 2433 | |||
BC158024.1 | - | 5 | 519 | 9088153 | ||
CK617936.1 | - | 5 | 559 | |||
D10917.1 | - | 1 | 65 |