Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017819.1 | - | 1 | 571 | ||
BC027431.1 | - | 1 | 439 | 3156359 | ||
CX235646.1 | - | 1 | 412 | |||
X56044.1 | - | 1 | 766 |