Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080999.2 | NP_001074468.1 | 2 | 881 | ||
NM_001081000.2 | NP_001074469.1 | 2 | 928 | |||
NM_001195205.1 | NP_001182134.1 | 2 | 881 | |||
GenBank mRNA | AK017819.1 | - | 1 | 1125 | ||
AK034102.1 | - | 2 | 761 | |||
AK080170.1 | - | 2 | 756 | |||
AK133844.1 | - | 2 | 756 | |||
AK134368.1 | - | 2 | 882 | |||
AK146391.1 | - | 2 | 821 | |||
AK150482.1 | - | 2 | 726 | |||
AK151311.1 | - | 2 | 726 | |||
AK151422.1 | - | 2 | 752 | |||
AK151725.1 | - | 2 | 726 | |||
AK151778.1 | - | 2 | 726 | |||
AK152055.1 | - | 2 | 726 | |||
AK152201.1 | - | 2 | 726 | |||
AK152363.1 | - | 2 | 726 | |||
AK152548.1 | - | 2 | 726 | |||
AK152903.1 | - | 2 | 726 | |||
AK153043.1 | - | 2 | 726 | |||
AK153190.1 | - | 2 | 752 | |||
AK169240.1 | - | 2 | 746 | |||
BC027431.1 | - | 1 | 993 | 3156359 | ||
BC038486.1 | - | 2 | 754 | 3155739 | ||
BC046976.1 | - | 2 | 762 | 6337243 | ||
X56044.1 | - | 1 | 1324 |