Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008313.4 | NP_032339.2 | 7 | 1433 | ||
GenBank mRNA | AK141587.1 | - | 7 | 1377 | ||
AK141625.1 | - | 7 | 1434 | |||
BC132189.1 | - | 6 | 1262 | |||
BC132191.1 | - | 6 | 1262 | |||
BC145159.1 | - | 6 | 1262 | |||
BC145161.1 | - | 7 | 1320 | |||
Y09585.1 | - | 6 | 1222 |
Set Membership: |
G-protein coupled receptor |