Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF076845.1 | - | 2, 3 | 203 | ||
AK046152.1 | - | 2, 3 | 213 | |||
AK051839.1 | - | 1, 2 | 638 | |||
AK088403.1 | - | 2, 3 | 213 | |||
AK135922.1 | - | 2, 3 | 193 | |||
BB611267.1 | - | 2, 3 | 269 | |||
BC047460.1 | - | 2, 3 | 220 | |||
BC061249.1 | - | 2, 3 | 239 | 30249148 | ||
Y16894.2 | - | 2, 3 | 235 |