Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293690.1 | NP_001280619.1 | 7 | 789 | ||
NM_008324.2 | NP_032350.1 | 7 | 636 | |||
GenBank mRNA | AK033783.1 | - | 7 | 790 | ||
BC049931.1 | - | 7 | 627 | 5387937 | ||
M69109.1 | - | 7 | 693 |