Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008325.4 | NP_032351.2 | 15 | 3434 | ||
NM_172997.1 | NP_766585.1 | 14 | 3293 | |||
GenBank mRNA | AK045330.1 | - | 13 | 2921 | ||
AK155627.1 | - | 15 | 3438 | |||
AK169998.1 | - | 14 | 3100 | |||
AK171124.1 | - | 10 | 2452 | |||
L34111.1 | - | 14 | 3005 |