Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008348.2 | NP_032374.1 | 5 | 751 | ||
GenBank mRNA | AK137663.1 | - | 5 | 752 | ||
AK149680.1 | - | 5 | 746 | |||
AK149899.1 | - | 4 | 634 | |||
AK150022.1 | - | 5 | 748 | |||
AK150524.1 | - | 5 | 755 | |||
AK153120.1 | - | 5 | 755 | |||
AK159566.1 | - | 5 | 748 | |||
L12120.1 | - | 5 | 756 |