Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008349.5 | NP_032375.2 | 5 | 674 | ||
GenBank mRNA | AF440787.1 | - | 5 | 601 | ||
AK040938.1 | - | 5 | 590 | |||
AK134042.1 | - | 5 | 667 | |||
AK160991.1 | - | 5 | 1629 | |||
AK181086.1 | - | 1 | 51 | |||
AK196209.1 | - | 2 | 161 | |||
AK201410.1 | - | 1 | 51 | |||
BC145791.1 | - | 5 | 674 | 40131094 | ||
U53696.1 | - | 5 | 587 |