Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF128214.1 | - | 7 | 981 | ||
AF128215.1 | - | 7 | 981 | |||
AK162981.1 | - | 7 | 1005 | |||
BC103608.1 | - | 7 | 987 | 40046200 | ||
BC103609.1 | - | 7 | 987 | 40046202 | ||
BC103610.1 | - | 7 | 987 | 40046204 | ||
BC103614.1 | - | 7 | 987 | 40046206 | ||
M86671.1 | - | 7 | 982 |