Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254747.1 | NP_001241676.1 | 8 | 1163 | ||
NM_008357.2 | NP_032383.1 | 8 | 1173 | |||
GenBank mRNA | AK155616.1 | - | 8 | 1811 | ||
BC023698.1 | - | 8 | 1158 | 5345382 | ||
U14332.1 | - | 8 | 1132 |