Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177973.1 | NP_001171444.1 | 15 | 2679 | ||
NM_001177975.1 | NP_001171446.1 | 14 | 2559 | |||
NM_008363.2 | NP_032389.2 | 14 | 2559 | |||
GenBank mRNA | AF103876.1 | - | 14 | 2559 | ||
AK045958.1 | - | 14 | 2468 | |||
AK052933.1 | - | 12 | 2429 | |||
AK143694.1 | - | 14 | 2470 | |||
AK144879.1 | - | 14 | 2468 | |||
AY184362.1 | - | 14 | 2559 | |||
BC004778.1 | - | 15 | 2671 | 3257579 | ||
U56773.2 | - | 14 | 2383 |