Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008366.3 | NP_032392.1 | 4 | 556 | ||
GenBank mRNA | AF065914.1 | - | 4 | 554 | ||
AF065915.1 | - | 4 | 554 | |||
AF065916.1 | - | 4 | 554 | |||
AF352786.1 | - | 1 | 35 | |||
BC116845.1 | - | 4 | 554 | 40126164 | ||
BC116873.1 | - | 4 | 554 | 40126192 | ||
BX519775.1 | - | 1 | 35 | |||
K02292.1 | - | 4 | 556 | |||
X01772.1 | - | 4 | 555 | |||
X73040.1 | - | 4 | 555 |