Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008369.1 | NP_032395.1 | 8, 9 | 981 | ||
GenBank mRNA | BC049889.1 | - | 7, 8 | 1381 | ||
FJ550346.1 | - | 5, 6 | 563 | |||
X64534.1 | - | 8, 9 | 981 |