Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008371.4 | NP_032397.1 | 5 | 708 | ||
GenBank mRNA | AK040399.1 | - | 5 | 492 | ||
AK041307.1 | - | 5 | 709 | |||
AK041403.1 | - | 410 | ||||
AK052452.1 | - | 6 | 1396 | |||
AK139858.1 | - | 6 | 1069 | |||
AK145184.1 | - | 5 | 999 | |||
BC110553.2 | - | 5 | 617 | 40047584 | ||
X07962.1 | - | 5 | 1008 |