Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008371.4 | NP_032397.1 | 5 | 1191 | ||
GenBank mRNA | AK040399.1 | - | 5 | 975 | ||
AK041307.1 | - | 5 | 1192 | |||
AK041403.1 | - | 893 | ||||
AK052452.1 | - | 6 | 1879 | |||
AK139858.1 | - | 6 | 1552 | |||
AK145184.1 | - | 5 | 1482 | |||
BC110553.2 | - | 5 | 1100 | 40047584 | ||
X07962.1 | - | 5 | 1491 |