Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008371.4 | NP_032397.1 | 5 | 1802 | ||
GenBank mRNA | AK040399.1 | - | 5 | 1586 | ||
AK041307.1 | - | 5 | 1804 | |||
AK041403.1 | - | 1504 | ||||
AK145184.1 | - | 5 | 2084 | |||
BC110553.2 | - | 5 | 1705 | 40047584 |