Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il9r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134458.1 | NP_001127930.1 | 9 | 2039 | ||
NM_008374.2 | NP_032400.1 | 9 | 2036 | |||
GenBank mRNA | M84746.1 | - | 9 | 1978 |