Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162425.1 | NP_001155897.1 | 5 | 597 | ||
NM_010107.4 | NP_034237.3 | 5 | 633 | |||
GenBank mRNA | AK009144.1 | - | 5 | 630 | ||
AK079266.1 | - | 5 | 600 | |||
BC002046.1 | - | 5 | 598 | 3484518 | ||
D38146.1 | - | 5 | 530 | |||
U26188.1 | - | 5 | 592 | |||
U90662.1 | - | 5 | 532 |