Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4ebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010124.2 | NP_034254.1 | 2 | 282 | ||
GenBank mRNA | AK133912.1 | - | 2 | 170 | ||
AK148374.1 | - | 321 | ||||
AK210066.1 | - | 1 | 20 | |||
AK211033.1 | - | 1 | 18 | |||
AK216468.1 | - | 1 | 20 | |||
AK217241.1 | - | 1 | 20 | |||
AK217805.1 | - | 1 | 20 | |||
BC015082.1 | - | 3 | 263 | 4218820 | ||
BC119539.2 | - | 3 | 246 | 40056467 | ||
U75530.1 | - | 2 | 279 |