Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4ebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010124.2 | NP_034254.1 | 3 | 1487 | ||
GenBank mRNA | AK133912.1 | - | 3 | 1374 | ||
AK147814.1 | - | 148 | ||||
AK148374.1 | - | 1528 | ||||
AK164011.1 | - | 422 | ||||
BC015082.1 | - | 4 | 1468 | 4218820 | ||
U75530.1 | - | 3 | 1482 |