Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010132.2 | NP_034262.2 | 3 | 2184 | ||
GenBank mRNA | AK132093.1 | - | 3 | 2018 | ||
AY117415.1 | - | 3 | 2548 |