Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083120.2 | NP_001076589.1 | 13, 14 | 2761 | ||
NM_001083121.2 | NP_001076590.1 | 13, 14 | 2029 | |||
NM_008680.3 | NP_032706.2 | 14, 15 | 2773 | |||
NM_010135.3 | NP_034265.2 | 14, 15 | 2818 | |||
GenBank mRNA | BC062927.1 | - | 13, 14 | 1648 | 6851217 | |
U72520.1 | - | 13, 14 | 1722 | |||
U72521.1 | - | 13, 14 | 2448 | |||
U72522.1 | - | 14, 15 | 2460 | |||
U72523.1 | - | 14, 15 | 2505 |