Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010140.3 | NP_034270.1 | 6 | 1433 | ||
GenBank mRNA | AK041935.1 | - | 6 | 1434 | ||
AK045219.1 | - | 6 | 1434 | |||
AK084921.1 | - | 6 | 1413 | |||
AK132335.1 | - | 6 | 1382 | |||
BC053479.1 | - | 2 | 325 | |||
BC060164.1 | - | 6 | 1410 | |||
BC093483.1 | - | 1434 | ||||
BC138641.1 | - | 6 | 1356 | 8861661 | ||
BC138643.1 | - | 6 | 1356 | 8861663 | ||
BC145349.1 | - | 6 | 1356 | 9053905 | ||
M68513.1 | - | 6 | 1408 | |||
M68515.1 | - | 6 | 1408 |