Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010145.2 | NP_034275.1 | 3 | 375 | ||
GenBank mRNA | AK018249.1 | - | 3 | 384 | ||
BC029636.1 | - | 3 | 374 | 3491966 | ||
BC045194.1 | - | 3 | 375 | 4986422 | ||
BC057857.1 | - | 3 | 370 | 5067895 | ||
BC061493.1 | - | 3 | 401 | 4488885 | ||
U89491.1 | - | 3 | 344 |