Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033711.1 | NP_001028883.1 | 2 | 1595 | ||
NM_010161.3 | NP_034291.1 | 2 | 782 | |||
GenBank mRNA | AK044098.1 | - | 2 | 658 | ||
AK046407.1 | - | 2 | 733 | |||
AK160302.1 | - | 2 | 699 | |||
AK162958.1 | - | 658 | ||||
AK170527.1 | - | 2 | 754 | |||
AK171332.1 | - | 1 | 3139 | |||
AK171594.1 | - | 2 | 700 | |||
BC038124.1 | - | 2 | 739 | 1396641 | ||
M34896.1 | - | 2 | 712 | |||
M34897.1 | - | 1 | 1108 |