Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252192.1 | NP_001239121.1 | 13 | 1803 | ||
NM_010164.2 | NP_034294.2 | 14 | 1919 | |||
GenBank mRNA | AJ007995.1 | - | 1 | 33 | ||
AK042412.1 | - | 13 | 1726 | |||
AK153788.1 | - | 14 | 1925 | |||
AK162111.1 | - | 6 | 660 | |||
BC060260.1 | - | 14 | 1919 | 6848408 | ||
BC066860.1 | - | 14 | 1573 | 30089692 | ||
U61110.1 | - | 13 | 1476 |