Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205243.1 | NP_001192172.1 | 3 | 718 | ||
NM_010177.4 | NP_034307.1 | 4 | 764 | |||
GenBank mRNA | AF288572.1 | - | 3 | 508 | ||
AK040139.1 | - | 4 | 764 | |||
AK141268.1 | - | 4 | 764 | |||
BC052866.1 | - | 32 | 30073084 | |||
S76752.1 | - | 4 | 584 | |||
U06948.1 | - | 4 | 678 | |||
U10984.1 | - | 4 | 734 |