Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005095.2 | NP_005086.2 | 7 | 1163 | ||
XM_005271328.4 | XP_005271385.1 | 7 | 1442 | |||
XM_005271331.2 | XP_005271388.1 | 4 | 408 | |||
XM_011542421.2 | XP_011540723.1 | 7 | 1089 | |||
XM_011542423.2 | XP_011540725.1 | 8 | 1494 | |||
XM_011542425.2 | XP_011540727.1 | 5 | 448 | |||
XM_017002803.1 | XP_016858292.1 | 8 | 1216 | |||
XM_017002804.1 | XP_016858293.1 | 8 | 1156 | |||
GenBank mRNA | AB007885.1 | - | 3 | 283 | ||
AK308320.1 | - | 7 | 1164 | |||
AK309705.1 | - | 6 | 823 | |||
BC012093.1 | - | 5 | 742 | |||
BC127113.1 | - | 8 | 1169 | 40125110 | ||
BC127114.1 | - | 4 | 352 | 40125115 | ||
BX647376.1 | - | 3 | 393 | |||
CR936741.1 | - | 6 | 1027 |