Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077189.1 | NP_001070657.1 | 5 | 779 | ||
NM_010187.2 | NP_034317.1 | 5 | 779 | |||
GenBank mRNA | AK080885.1 | - | 5 | 782 | ||
AK089266.1 | - | 5 | 780 | |||
AK150613.1 | - | 5 | 780 | |||
AK155329.1 | - | 5 | 771 | |||
AK170035.1 | - | 5 | 769 | |||
AK171254.1 | - | 5 | 771 | |||
AK171316.1 | - | 5 | 762 | |||
BC011305.1 | - | 5 | 742 | |||
BC019758.1 | - | 5 | 742 | |||
BC038070.1 | - | 5 | 766 | 3993915 | ||
M14216.1 | - | 5 | 737 | |||
M16367.1 | - | 7 | 1020 | |||
M17515.1 | - | 5 | 733 | |||
U31801.1 | - | 3 | 594 | |||
U31802.1 | - | 3 | 594 | |||
U31803.1 | - | 3 | 594 | |||
U31804.1 | - | 3 | 594 | |||
U51629.1 | - | 5 | 687 |