Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077189.1 | NP_001070657.1 | 8 | 1075 | ||
NM_010187.2 | NP_034317.1 | 7 | 934 | |||
GenBank mRNA | AK089266.1 | - | 7 | 935 | ||
AK150613.1 | - | 7 | 935 | |||
AK155329.1 | - | 8 | 1067 | |||
AK170035.1 | - | 7 | 924 | |||
AK171254.1 | - | 8 | 1067 | |||
AK171316.1 | - | 8 | 1058 | |||
BC011305.1 | - | 7 | 897 | |||
BC019758.1 | - | 7 | 897 | |||
BC038070.1 | - | 7 | 921 | 3993915 | ||
M14216.1 | - | 7 | 892 | |||
M16367.1 | - | 10 | 1313 | |||
U31801.1 | - | 6 | 890 | |||
U31802.1 | - | 6 | 890 | |||
U31803.1 | - | 5 | 749 | |||
U31804.1 | - | 5 | 749 | |||
U51629.1 | - | 8 | 899 |