Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PXK SIRNA Assays ›
Gene Symbol:
PXK
Gene Name:
PX domain containing serine/threonine kinase like
Gene Aliases:
MONaKA
Chromosome Location:
Chr.3: 58332868 - 58426127 on Build GRCh38
UniGene ID:
Hs.190544
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1585
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PXK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289095.1
NP_001276024.1
3
264
NM_001289096.1
NP_001276025.1
2
216
NM_001289098.1
NP_001276027.1
4
315
NM_001289099.1
NP_001276028.1
4
420
NM_001289100.1
NP_001276029.1
2
216
NM_017771.4
NP_060241.2
4
315
XM_005265250.4
XP_005265307.1
4
315
XM_005265252.4
XP_005265309.1
4
315
XM_005265255.3
XP_005265312.1
4
315
XM_005265256.3
XP_005265313.1
4
315
XM_017006671.1
XP_016862160.1
3
206
XM_017006672.1
XP_016862161.1
3
206
XM_017006673.1
XP_016862162.1
2
207
XM_017006674.1
XP_016862163.1
3
206
XM_017006675.1
XP_016862164.1
4
315
XM_017006676.1
XP_016862165.1
4
315
XM_017006677.1
XP_016862166.1
4
315
XM_017006678.1
XP_016862167.1
4
315
XM_017006679.1
XP_016862168.1
4
315
XM_017006680.1
XP_016862169.1
4
315
XM_017006681.1
XP_016862170.1
3
206
XM_017006682.1
XP_016862171.1
3
206
XM_017006683.1
XP_016862172.1
3
206
XM_017006684.1
XP_016862173.1
3
206
XM_017006685.1
XP_016862174.1
3
206
XM_017006686.1
XP_016862175.1
5
429
XM_017006687.1
XP_016862176.1
5
427
XM_017006688.1
XP_016862177.1
2
191
XM_017006689.1
XP_016862178.1
2
181
XM_017006690.1
XP_016862179.1
2
172
XM_017006692.1
XP_016862181.1
5
466
XM_017006693.1
XP_016862182.1
5
466
XM_017006694.1
XP_016862183.1
5
360
XM_017006695.1
XP_016862184.1
5
466
GenBank mRNA
AB209688.1
-
4
294
AF399753.1
-
4
216
AK000342.1
-
4
313
AK131385.1
-
3
206
AK295542.1
-
2
208
AY274811.1
-
4
315
AY437879.1
-
2
159
AY847220.1
-
4
323
AY847222.1
-
4
218
BC008943.2
-
4
296
BC014479.1
-
4
231
4856139
DA296189.1
-
4
316
DQ124707.1
-
4
216
DQ124708.1
-
4
216
HY164612.1
-
4
383
KF774202.1
-
4
325
KF774203.1
-
4
325
KF774204.1
-
4
400
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
inflammatory response
negative regulation of ATPase activity
regulation of membrane potential
negative regulation of ion transport
modulation of synaptic transmission
nucleotide binding
actin binding
protein kinase activity
ATP binding
protein C-terminus binding
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline