Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076062.1 | - | 3 | 423 | ||
AK136159.1 | - | 5 | 1204 | |||
AK146766.1 | - | 3 | 431 | |||
AK158633.1 | - | 3 | 420 | |||
AK162294.1 | - | 3 | 384 | |||
AK168629.1 | - | 3 | 357 | |||
AK169317.1 | - | 3 | 436 | |||
BC054722.1 | - | 3 | 365 | 5694816 | ||
BC138301.1 | - | 3 | 419 | 8861320 | ||
BC138302.1 | - | 3 | 420 | 8861321 | ||
D29016.1 | - | 3 | 373 |