Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145858.2 | NP_665857.2 | 3 | 275 | ||
GenBank mRNA | AF029689.1 | - | 3 | 142 | ||
AF098798.1 | - | 3 | 142 | |||
AK001293.1 | - | 3 | 168 | |||
AK057604.1 | - | 3 | 759 | |||
AK315707.1 | - | 3 | 135 | |||
BC013155.1 | - | 3 | 143 | |||
BC033023.2 | - | 3 | 129 | 5269248 | ||
BI599245.1 | - | 3 | 180 | |||
BX647276.1 | - | 3 | 162 | |||
BX648547.1 | - | 4 | 289 | |||
DB037192.1 | - | 3 | 276 |