Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010219.3 | NP_034349.1 | 5, 6 | 796 | ||
GenBank mRNA | AK051433.1 | - | 822 | |||
AK083912.1 | - | 5, 6 | 796 | |||
AK089972.1 | - | 5, 6 | 805 | |||
AK159211.1 | - | 5, 6 | 823 | |||
AK160202.1 | - | 5, 6 | 745 | |||
AK181993.1 | - | 1, 2 | 120 | |||
AK188096.1 | - | 1, 2 | 120 | |||
BC003447.1 | - | 5, 6 | 813 | 2651490 | ||
X17069.2 | - | 5, 6 | 661 | |||
X70887.1 | - | 5, 6 | 693 |