Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163481.1 | NP_001156953.1 | 3 | 511 | ||
NM_010221.2 | NP_034351.2 | 3 | 511 | |||
GenBank mRNA | AK019446.2 | - | 3 | 449 | ||
AK144185.1 | - | 3 | 494 | |||
BC029546.1 | - | 3 | 458 | 5059670 | ||
L07063.1 | - | 3 | 514 |