Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010229.2 | NP_034359.2 | 6 | 892 | ||
GenBank mRNA | AK045865.1 | - | 6 | 715 | ||
AK149292.1 | - | 6 | 710 | |||
AK163640.1 | - | 6 | 893 | |||
BC109003.1 | - | 6 | 685 | |||
BC109004.1 | - | 6 | 697 | |||
M64689.1 | - | 6 | 698 | |||
X59398.1 | - | 6 | 749 |