Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010231.2 | NP_034361.1 | 9 | 2126 | ||
GenBank mRNA | BC011229.1 | - | 9 | 2126 | 4196446 | |
D16215.1 | - | 9 | 2016 | |||
U87456.1 | - | 9 | 2111 |